Anti-FCER1A Mouse Monoclonal Antibody [clone: 9E1]

Supplier: GeneTex
GTX41157
89333-716EA 255.69 USD
89333-716
Anti-FCER1A Mouse Monoclonal Antibody [clone: 9E1]
Antibodies
Mouse monoclonal antibody [9E1] to Fc epsilon R1 alpha

Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: 9E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR