Anti-Feline Immunodeficiency Virus gp95 Mouse Monoclonal Antibody [clone: SU1-12]

GTX41154
89275-476EA 494.9 USD
89275-476
Anti-Feline Immunodeficiency Virus gp95 Mouse Monoclonal Antibody [clone: SU1-12]
Type: Primary
Antigen: Feline Immunodeficiency Virus gp95
Clonality: Monoclonal
Clone: SU1-12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR