Anti-Feline Immunodeficiency Virus gp95 Mouse Monoclonal Antibody [clone: SU2-3]

GTX41152
89275-472EA 494.9 USD
89275-472
Anti-Feline Immunodeficiency Virus gp95 Mouse Monoclonal Antibody [clone: SU2-3]
Type: Primary
Antigen: Feline Immunodeficiency Virus gp95
Clonality: Monoclonal
Clone: SU2-3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR