Anti-Feline Immunodeficiency Virus P24 Gag Mouse Monoclonal Antibody [clone: P24CR1]

GTX41150
89274-914EA 592.26 USD
89274-914
Anti-Feline Immunodeficiency Virus P24 Gag Mouse Monoclonal Antibody [clone: P24CR1]
Type: Primary
Antigen: Feline Immunodeficiency Virus P24 Gag
Clonality: Monoclonal
Clone: P24CR1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR