Anti-Feline Leukaemia Virus P27 Mouse Monoclonal Antibody [clone: PF12J-10A]

Supplier: GeneTex
GTX41145
89274-906EA 796.44 USD
89274-906
Anti-Feline Leukaemia Virus P27 Mouse Monoclonal Antibody [clone: PF12J-10A]
Antibodies
Type: Primary
Antigen: Feline Leukaemia Virus P27
Clonality: Monoclonal
Clone: PF12J-10A
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR