Anti-FTH1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX41144
89276-296EA 582.3 USD
89276-296
Anti-FTH1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Ferritin

Type: Primary
Antigen: Fth1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR