Anti-Streptococcus agalactiae Mouse Monoclonal Antibody [clone: BDI560]

Supplier: GeneTex
GTX41126
89336-946EA 659.34 USD
89336-946
Anti-Streptococcus agalactiae Mouse Monoclonal Antibody [clone: BDI560]
Antibodies
Type: Primary
Antigen: Streptococcus agalactiae
Clonality: Monoclonal
Clone: BDI560
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR