Anti-Influenza A Hemagglutinin H5 (Avian H5N1) Mouse Monoclonal Antibody [clone: B256M]

Supplier: GeneTex
GTX41120
89316-752EA 622.53 USD
89316-752
Anti-Influenza A Hemagglutinin H5 (Avian H5N1) Mouse Monoclonal Antibody [clone: B256M]
Antibodies
Type: Primary
Antigen: Influenza A Hemagglutinin H5 (Avian H5N1)
Clonality: Monoclonal
Clone: B256M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR