Anti-FST Mouse Monoclonal Antibody [clone: 17/2]

Supplier: GeneTex
GTX41094
89262-480EA 915.37 USD
89262-480
Anti-FST Mouse Monoclonal Antibody [clone: 17/2]
Mouse monoclonal antibody [17/2] to Follistatin 288

Type: Primary
Antigen: FST
Clonality: Monoclonal
Clone: 17/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR