Anti-FOXP1 Mouse Monoclonal Antibody [clone: JC12]

Supplier: GeneTex
GTX41073
89277-500EA 535.5 USD
89277-500
Anti-FOXP1 Mouse Monoclonal Antibody [clone: JC12]
Antibodies
Mouse monoclonal antibody [JC12] to FOXP1

Recommended Starting Dilutions:Immunohistology - Paraffin: 1/200 - 1/500Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: FOXP1
Clonality: Monoclonal
Clone: JC12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR