Anti-Influenza A Hemagglutinin H7 Mouse Monoclonal Antibody [clone: B1275M]

Supplier: GeneTex
GTX41071
89316-736EA 635.87 USD
89316-736
Anti-Influenza A Hemagglutinin H7 Mouse Monoclonal Antibody [clone: B1275M]
Antibodies
Type: Primary
Antigen: Influenza A Hemagglutinin H7
Clonality: Monoclonal
Clone: B1275M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR