Anti-FSH Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX41063
89265-046EA 765.28 USD
89265-046
Anti-FSH Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to FSH

Recommended Starting Dilutions:Immunohistology - Frozen: 1/200 - 1/500Western Blotting: 1/300 - 1/400Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR