Anti-Influenza A Hemagglutinin H9 Mouse Monoclonal Antibody [clone: B1286M]

GTX41056
89295-194EA 405.93 USD
89295-194
Anti-Influenza A Hemagglutinin H9 Mouse Monoclonal Antibody [clone: B1286M]
Type: Primary
Antigen: Influenza A Hemagglutinin H9
Clonality: Monoclonal
Clone: B1286M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR