Anti-Influenza A Nonstructural Protein (NS1) Mouse Monoclonal Antibody [clone: B467M]

Supplier: GeneTex
GTX41054
89295-190EA 611.89 USD
89295-190
Anti-Influenza A Nonstructural Protein (NS1) Mouse Monoclonal Antibody [clone: B467M]
Antibodies
Type: Primary
Antigen: Influenza A Nonstructural Protein (NS1)
Clonality: Monoclonal
Clone: B467M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR