Anti-GABRA6 Rabbit Polyclonal Antibody

GTX41048
89276-420EA 696.91 USD
89276-420
Anti-GABRA6 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to GABA A Receptor alpha 6

Recommended Starting Dilutions:Western Blotting: Max Dilution 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Gabra6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR