Anti-Influenza B (matrix protein M1) Mouse Monoclonal Antibody [clone: B278M]

Supplier: GeneTex
GTX41043
89299-604EA 448.34 USD
89299-604
Anti-Influenza B (matrix protein M1) Mouse Monoclonal Antibody [clone: B278M]
Antibodies
Type: Primary
Antigen: Influenza B (matrix protein M1)
Clonality: Monoclonal
Clone: B278M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR