Anti-Influenza B (Nucleoprotein) Mouse Monoclonal Antibody [clone: B265M]

Supplier: GeneTex
GTX41040
89278-588EA 640.93 USD
89278-588
Anti-Influenza B (Nucleoprotein) Mouse Monoclonal Antibody [clone: B265M]
Antibodies
Type: Primary
Antigen: Influenza B (Nucleoprotein)
Clonality: Monoclonal
Clone: B265M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR