Anti-Influenza B (Nucleoprotein) Mouse Monoclonal Antibody [clone: B267M]

Supplier: GeneTex
GTX41036
89278-586EA 640.74 USD
89278-586
Anti-Influenza B (Nucleoprotein) Mouse Monoclonal Antibody [clone: B267M]
Antibodies
Type: Primary
Antigen: Influenza B (Nucleoprotein)
Clonality: Monoclonal
Clone: B267M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR