Anti-BMP15 Mouse Monoclonal Antibody [clone: mAb-GDF9-53]

Supplier: GeneTex
GTX41002
89262-468EA 927.61 USD
89262-468
Anti-BMP15 Mouse Monoclonal Antibody [clone: mAb-GDF9-53]
Type: Primary
Antigen: BMP15
Clonality: Monoclonal
Clone: mAb-GDF9-53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR