Anti-HBcAg Mouse Monoclonal Antibody [clone: H50]

Supplier: GeneTex
GTX40726
89265-888EA 656.38 USD
89265-888
Anti-HBcAg Mouse Monoclonal Antibody [clone: H50]
Antibodies
Type: Primary
Antigen: Hepatitis B Core Antigen
Clonality: Monoclonal
Clone: H50
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR