Anti-HBsAg Mouse Monoclonal Antibody [clone: 1834]

Supplier: GeneTex
GTX40707
89280-528EA 662.45 USD
89280-528
Anti-HBsAg Mouse Monoclonal Antibody [clone: 1834]
Antibodies
Type: Primary
Antigen: Hepatitis B Surface Antigen
Clonality: Monoclonal
Clone: 1834
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR