Anti-Hepatitis C Core Antigen Mouse Monoclonal Antibody [clone: 1868]

Supplier: GeneTex
GTX40705
89275-444EA 738.87 USD
89275-444
Anti-Hepatitis C Core Antigen Mouse Monoclonal Antibody [clone: 1868]
Antibodies
Type: Primary
Antigen: Hepatitis C Core Antigen
Clonality: Monoclonal
Clone: 1868
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR