Anti-Hepatitis C E1 Antigen Mouse Monoclonal Antibody [clone: 1879]

GTX40702
89276-408EA 696.3 USD
89276-408
Anti-Hepatitis C E1 Antigen Mouse Monoclonal Antibody [clone: 1879]
Type: Primary
Antigen: Hepatitis C E1 Antigen
Clonality: Monoclonal
Clone: 1879
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR