Anti-Klebsiella pneumoniae 204 Mouse Monoclonal Antibody [clone: B394M]

GTX40674
89295-184EA 403.33 USD
89295-184
Anti-Klebsiella pneumoniae 204 Mouse Monoclonal Antibody [clone: B394M]
Type: Primary
Antigen: Klebsiella pneumoniae 204
Clonality: Monoclonal
Clone: B394M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR