Anti-Hepatitis C NS-5 Antigen Mouse Monoclonal Antibody [clone: 5B7 (BGN/1246/5B7)]

GTX40673
89275-432EA 494.9 USD
89275-432
Anti-Hepatitis C NS-5 Antigen Mouse Monoclonal Antibody [clone: 5B7 (BGN/1246/5B7)]
Type: Primary
Antigen: Hepatitis C NS-5 Antigen
Clonality: Monoclonal
Clone: 5B7 (BGN/1246/5B7)
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR