Anti-HSV2 Mouse Monoclonal Antibody [clone: 1073/53]

GTX40660
89265-884EA 451 USD
89265-884
Anti-HSV2 Mouse Monoclonal Antibody [clone: 1073/53]
Type: Primary
Antigen: HSV2
Clonality: Monoclonal
Clone: 1073/53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR