Anti-LMNB2 Mouse Monoclonal Antibody [clone: X167]

GTX40642
89282-148EA 378.69 USD
89282-148
Anti-LMNB2 Mouse Monoclonal Antibody [clone: X167]
Mouse Monoclonal antibody [X167] to Lamin

Type: Primary
Antigen: LMNB2
Clonality: Monoclonal
Clone: X167
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR