Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 7711]

Supplier: GeneTex
GTX40639
89288-242EA 478.51 USD
89288-242
Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 7711]
Antibodies
Type: Primary
Antigen: Campylobacter jejuni
Clonality: Monoclonal
Clone: 7711
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR