Anti-HLA ABC Mouse Monoclonal Antibody [clone: Bu8]

GTX40614
89333-706EA 199.73 USD
89333-706
Anti-HLA ABC Mouse Monoclonal Antibody [clone: Bu8]
Mouse monoclonal antibody [Bu8] to HLA-ABC

Recommended Starting Dilutions:Flow Cytometry: 1/25 - 1/50Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HLA ABC
Clonality: Monoclonal
Clone: Bu8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR