Anti-Legionella pneumophila LPS Mouse Monoclonal Antibody [clone: B1252M]

Supplier: GeneTex
GTX40593
89282-794EA 818.81 USD
89282-794
Anti-Legionella pneumophila LPS Mouse Monoclonal Antibody [clone: B1252M]
Antibodies
Type: Primary
Antigen: Legionella pneumophila LPS
Clonality: Monoclonal
Clone: B1252M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR