Anti-HIV1 p24 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX40569
89337-040EA 659.34 USD
89337-040
Anti-HIV1 p24 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat polyclonal antibody to HIV1 p24 (FITC)

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Direct FA staining of target antigens in a permissive tissue culture system. Working dilution must be determined by the user but a starting range of 1:10 - 1:50 is suggested. Acetone fixation of the antigen source is recommended prior to staining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HIV1 p24
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR