Anti-Homing Receptor Mouse Monoclonal Antibody [clone: CT4]

Supplier: GeneTex
GTX40566
89275-966EA 638.11 USD
89275-966
Anti-Homing Receptor Mouse Monoclonal Antibody [clone: CT4]
Antibodies
Type: Primary
Antigen: Homing Receptor
Clonality: Monoclonal
Clone: CT4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR