Anti-LEP Mouse Monoclonal Antibody [clone: BDI142]

GTX40565
89286-968EA 403.61 USD
89286-968
Anti-LEP Mouse Monoclonal Antibody [clone: BDI142]
Type: Primary
Antigen: LEP
Clonality: Monoclonal
Clone: BDI142
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR