Anti-ALDOA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GeneTex
GTX40552
89336-726EA 762.27 USD
89336-726
Anti-ALDOA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: ALDOA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR