Anti-HK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GeneTex
GTX40547
89336-722EA 762.27 USD
89336-722
Anti-HK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Peroxidase-conjugated Rabbit Polyclonal antibody to Hexokinase

Recommended Starting Dilutions:WB: Use at 1:500 - 1:2,000ELISA: Use at 1:4,000 - 1:20,000

Type: Primary
Antigen: HK1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR