Anti-Chlamydia trachomatis Goat Polyclonal Antibody (Biotin)

GTX40496
89287-812EA 399.48 USD
89287-812
Anti-Chlamydia trachomatis Goat Polyclonal Antibody (Biotin)
Type: Primary
Antigen: Chlamydia trachomatis
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR