Anti-Lymphocytic Choriomeningitis Virus (LCMV) Mouse Monoclonal Antibody [clone: M 104]

Supplier: GeneTex
GTX40495
89280-394EA 902.73 USD
89280-394
Anti-Lymphocytic Choriomeningitis Virus (LCMV) Mouse Monoclonal Antibody [clone: M 104]
Antibodies
Type: Primary
Antigen: Lymphocytic Choriomeningitis Virus (LCMV)
Clonality: Monoclonal
Clone: M 104
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR