Anti-IgA Mouse Monoclonal Antibody [clone: IgA5-3B]

Supplier: GeneTex
GTX40480
89274-892EA 766.32 USD
89274-892
Anti-IgA Mouse Monoclonal Antibody [clone: IgA5-3B]
Antibodies
Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: IgA5-3B
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Cat Dog, Monkey, Rhesus Monkey, Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR