Anti-IgA Mouse Monoclonal Antibody [clone: IgA9-6A]

Supplier: GeneTex
GTX40479
89275-418EA 646.52 USD
89275-418
Anti-IgA Mouse Monoclonal Antibody [clone: IgA9-6A]
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: IgA9-6A
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Cat, Dog, Rhesus Monkey, Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR