Anti-F8 Mouse Monoclonal Antibody [clone: 102]

Supplier: GeneTex
GTX40474
89337-038EA 659.34 USD
89337-038
Anti-F8 Mouse Monoclonal Antibody [clone: 102]
Antibodies
Mouse monoclonal [102] to Factor VIII

IA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR