Anti-PCP Mouse Monoclonal Antibody [clone: BDI970]

GTX40457
89286-450EA 376.58 USD
89286-450
Anti-PCP Mouse Monoclonal Antibody [clone: BDI970]
Type: Primary
Antigen: PCP
Clonality: Monoclonal
Clone: BDI970
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR