Anti-Duck IgA2 Mouse Monoclonal Antibody [clone: DuIgA2]

Supplier: GeneTex
GTX40454
89275-412EA 644.18 USD
89275-412
Anti-Duck IgA2 Mouse Monoclonal Antibody [clone: DuIgA2]
Antibodies
Type: Secondary
Antigen: IgA2
Clonality: Monoclonal
Clone: DuIgA2
Conjugation:
Epitope: Heavy chain
Host: Mouse
Isotype:
Reactivity: Duck
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR