Anti-IgD F(c) Mouse Monoclonal Antibody [clone: LE2]

Supplier: GeneTex
GTX40451
89265-878EA 611.26 USD
89265-878
Anti-IgD F(c) Mouse Monoclonal Antibody [clone: LE2]
Antibodies
Mouse Monoclonal antibody [LE2] to Human IgD (Fc)

Recommended Starting Dilutions:For IHC-Fr: Use at 1/100 - 1/300Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgD F(c)
Clonality: Monoclonal
Clone: LE2
Conjugation:
Epitope: F(c)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR