Anti-Major Basic Protein Mouse Monoclonal Antibody [clone: BMK13]

Supplier: GeneTex
GTX40448
89288-240EA 535.39 USD
89288-240
Anti-Major Basic Protein Mouse Monoclonal Antibody [clone: BMK13]
Antibodies
Type: Primary
Antigen: Major Basic Protein
Clonality: Monoclonal
Clone: BMK13
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR