Anti-Clostridium Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX40447
89337-036EA 659.34 USD
89337-036
Anti-Clostridium Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Clostridium
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR