Anti-Marburg Virus Mouse Monoclonal Antibody [clone: B438M]

Supplier: GeneTex
GTX40445
89282-782EA 545.66 USD
89282-782
Anti-Marburg Virus Mouse Monoclonal Antibody [clone: B438M]
Antibodies
Type: Primary
Antigen: Marburg Virus
Clonality: Monoclonal
Clone: B438M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR