Anti-IgE Mouse Monoclonal Antibody [clone: E6-71A1]

Supplier: GeneTex
GTX40438
89275-408EA 637.39 USD
89275-408
Anti-IgE Mouse Monoclonal Antibody [clone: E6-71A1]
Antibodies
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: E6-71A1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Cat Dog, Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR