Anti-MBP Mouse Monoclonal Antibody [clone: BDI221]

Supplier: GeneTex
GTX40406
89299-568EA 601.01 USD
89299-568
Anti-MBP Mouse Monoclonal Antibody [clone: BDI221]
Antibodies
Mouse Monoclonal antibody [BDI221] to MBP

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone: BDI221
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR