Anti-Methadone Mouse Monoclonal Antibody [clone: BID902]

Supplier: GeneTex
GTX40368
89299-562EA 611.43 USD
89299-562
Anti-Methadone Mouse Monoclonal Antibody [clone: BID902]
Antibodies
Type: Primary
Antigen: Methadone
Clonality: Monoclonal
Clone: BID902
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: [delete]
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR