Anti-IgG Mouse Monoclonal Antibody [clone: HP6200]

Supplier: GeneTex
GTX40349
89265-874EA 611.26 USD
89265-874
Anti-IgG Mouse Monoclonal Antibody [clone: HP6200]
Antibodies
Mouse Monoclonal antibody [HP6200] to Human IgG2 F(Ab)2'

Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: HP6200
Conjugation:
Epitope: F(ab')2
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR